• USA Home
  • 8.14077 - 4-Methoxyphenylhydrazinium chloride

EMAIL THIS PAGE TO A FRIEND
8.14077 EMD Millipore

4-Methoxyphenylhydrazinium chloride

4-Methoxyphenylhydrazinium chloride for synthesis. CAS 19501-58-7, chemical formula 4-(CH₃O)C₆H₄NHNH₂HCl., for synthesis

  • CAS Number 19501-58-7

  • Linear Formula C₇H₁₁ClN₂O

  • Molecular Weight 174.63 g/mol

  •  EC Number 243-115-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8140770010)
mp   158 - 161 °C decomposes
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 99.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2928 00 90
Hazard Statement(s) 
H315: Causes skin irritation.
Precautionary Statement(s) 
P262: Do not get in eyes, on skin, or on clothing.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 2811 , 6.1, I
Declaration (transport by air) IATA-DGR 
UN 2811 , 6.1, I
Declaration (transport by sea) IMDG-Code 
UN 2811 , 6.1, I
Categories of danger 
irritant
R Phrase 
R 38
Irritating to skin.
S Phrase 
S 22-25
Do not breathe dust.Avoid contact with eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?