• USA Home
  • 8.14083 - 4-tert-Butylcyclohexyl acetate

EMAIL THIS PAGE TO A FRIEND
8.14083 EMD Millipore

4-tert-Butylcyclohexyl acetate

4-tert-Butylcyclohexyl acetate (mixture of cis- and trans isomers) for synthesis. CAS 32210-23-4, chemical formula 4-(CH₃)₃C-C₆H₁₀-OOCCH₃., (mixture of cis- and trans isomers) for synthesis

Synonym: Acetic acid 4-tert-butylcyclohexyl ester

  • CAS Number 32210-23-4

  • Linear Formula C₁₂H₂₂O₂

  • Molecular Weight 198.3 g/mol

  •  EC Number 250-954-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
solubility   <0.0396 g/l insoluble
density   0.936 g/cm3 (20 °C)
flash point   104 °C
autoignition temp.   390 °C
viscosity kinematic   8.37 mm2/s (20 °C)
LD 50 oral   LD50 Rat 5000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.936 - 0.938
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2915 39 00
Hazard Statement(s) 
H317: May cause an allergic skin reaction.
H411: Toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P280: Wear protective gloves.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Irritant  Irritant
R Phrase 
R 43
May cause sensitisation by skin contact.
S Phrase 
S 24-37
Avoid contact with skin.Wear suitable gloves.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?