• USA Home
  • 8.14088 - Tributylmethylammonium chloride

EMAIL THIS PAGE TO A FRIEND
8.14088 EMD Millipore

Tributylmethylammonium chloride

(75% solution in water) for synthesis

Synonym: Methyltributylammonium chloride

Purchase

Properties

packaging   500 ml in Glass bottle (8140880500)
bp   84 - 85 °C (133 hPa)
vapor pressure   7.9 hPa (25 °C)
density   0.96 g/cm3 (20 °C)
mp   -33 °C
pH   8 (H₂O, 20 °C)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): 74.0 - 79.0 %
  Density (d 20 °C/ 4 °C): 0.960 - 0.970

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2923 90 00
Hazard Statement(s) 
H302 + H312: Harmful if swallowed or in contact with skin
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful, irritant
R Phrase 
R 21/22-36/37/38
Harmful in contact with skin and if swallowed.Irritating to eyes, respiratory system and skin.
S Phrase 
S 36/37
Wear suitable protective clothing and gloves.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?