• USA Home
  • 8.14089 - 4-Fluoro-3-nitroaniline

EMAIL THIS PAGE TO A FRIEND
8.14089 EMD Millipore

4-Fluoro-3-nitroaniline

4-Fluoro-3-nitroaniline for synthesis. CAS 364-76-1, molar mass 156.12 g/mol., for synthesis

  • CAS Number 364-76-1

  • Linear Formula C₆H₅FN₂O₂

  • Molecular Weight 156.12 g/mol

  •  EC Number 206-665-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8140890010)
density   0.43 g/cm3 (20 °C)
flash point   91 °C
mp   93 - 96 °C
LD 50 oral   LD50 Rat 1100 mg/kg
LD 50 dermal   LD50 Rabbit > 10000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 93 °C
  Melting range (upper value): ≤ 96 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 42 00
Hazard Statement(s) 
H302: Harmful if swallowed.
Precautionary Statement(s) 
P262: Do not get in eyes, on skin, or on clothing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
S Phrase 
S 24/25
Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?