• USA Home
  • 8.14094 - 4-Fluorophenylacetic acid

EMAIL THIS PAGE TO A FRIEND
8.14094 EMD Millipore

4-Fluorophenylacetic acid

4-Fluorophenylacetic acid for synthesis. CAS 405-50-5, chemical formula 4(F) * C₆H₄CH₂COOH., for synthesis

  • CAS Number 405-50-5

  • Linear Formula C₈H₇FO₂

  • Molecular Weight 154.14 g/mol

  •  EC Number 206-972-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8140940010)
  50 g in Glass bottle (8140940050)
bp   164 °C (3 hPa)
mp   81 - 84 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 98.0 %
  Melting range (lower value): ≥ 81 °C
  Melting range (upper value): ≤ 84 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 39 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?