• USA Home
  • 8.14153 - Dimethyl acetylsuccinate

EMAIL THIS PAGE TO A FRIEND
8.14153 EMD Millipore

Dimethyl acetylsuccinate

Dimethyl acetylsuccinate for synthesis. CAS 10420-33-4, chemical formula CH₃OOCCH₂CH(COCH₃)COOCH₃., for synthesis

Synonym: Acetylsuccinic acid dimethyl ester

  • CAS Number 10420-33-4

  • Linear Formula C₈H₁₂O₅

  • Molecular Weight 188.18 g/mol

  •  EC Number 233-897-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Plastic bottle (8141530100)
bp   248 °C (981 hPa)
expl. lim.     0.72 - 11.1 %(V)
vapor pressure   <1 hPa (20 °C)
solubility   30 g/l OECD Test Guideline 105
density   1.16 g/cm3 (20 °C) OECD Test Guideline 109
flash point   128 °C
autoignition temp.   340 °C DIN 51794
mp   28 - 31 °C
LD 50 oral   LD50 Rat > 2000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 95.0 % (a/a)
  Melting range (lower value): ≥ 28 °C
  Melting range (upper value): ≤ 31 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 30 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?