• USA Home
  • 8.14163 - 2-Methoxy-4-methylphenol

EMAIL THIS PAGE TO A FRIEND
8.14163 EMD Millipore

2-Methoxy-4-methylphenol

2-Methoxy-4-methylphenol for synthesis. CAS 93-51-6, chemical formula 2(CH₃O)-4-(CH₃)C₆H₃OH., for synthesis

Synonym: 2-Hydroxy-5-methylanisole, 4-Methylguajacol

  • CAS Number 93-51-6

  • Linear Formula C₈H₁₀O₂

  • Molecular Weight 138.17 g/mol

  •  EC Number 202-252-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8141630010)
bp   78 - 79 °C (5.3 hPa)
vapor pressure   0.032 hPa (25 °C)
density   1.09 g/cm3
flash point   85 °C
mp   5 - 6 °C
LD 50 oral   LD50 Rat 740 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.093 - 1.095
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 50 00
Hazard Statement(s) 
H302: Harmful if swallowed.
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
1
Strongly contaminated halogen-free organic solvents: container A.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful, irritant
R Phrase 
R 22-36/38
Harmful if swallowed.Irritating to eyes and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?