• USA Home
  • 8.14177 - Diisoheptyl phthalate

EMAIL THIS PAGE TO A FRIEND
8.14177 EMD Millipore

Diisoheptyl phthalate

Diisoheptyl phthalate for synthesis. CAS 71888-89-6, molar mass 362.51 g/mol., for synthesis

Synonym: Phthalic acid diisoheptyl ester

  • CAS Number 71888-89-6

  • Linear Formula C₂₂H₃₄O₄

  • Molecular Weight 362.51 g/mol

  •  EC Number 276-158-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8141770100)
bp   >300 °C (0.01 hPa)
vapor pressure   <0.10 hPa (20 °C)
solubility   1 g/l
density   0.99 g/cm3 (20 °C)
flash point   229 °C
autoignition temp.   380 °C
mp   -40 °C
LD 50 oral   LD50 Rat > 10000 mg/kg
LD 50 dermal   LD50 Rabbit > 3160 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 99.0 %
  Density (d 20 °C/ 4 °C): 0.991 - 0.993
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2917 34 00
Hazard Statement(s) 
H360D: May damage the unborn child.
Precautionary Statement(s) 
P201: Obtain special instructions before use.
P308 + P313: IF exposed or concerned: Get medical advice/ attention.
RTECS 
CZ3242500
Storage class 
6.1A Combustible substances, toxic
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS08  GHS08
Hazard Symbols 
Toxic  Toxic
Categories of danger 
toxic for reproduction
R Phrase 
R 61
May cause harm to the unborn child.
S Phrase 
S 53-45
Avoid exposure - obtain special instructions before use.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?