• USA Home
  • 8.14182 - 3-Amino-5-methylisoxazol

EMAIL THIS PAGE TO A FRIEND
8.14182 EMD Millipore

3-Amino-5-methylisoxazol

3-Amino-5-methylisoxazol for synthesis. CAS 1072-67-9, pH 6.0 - 7.5 (10 g/l, H₂O, 20 °C)., for synthesis

  • CAS Number 1072-67-9

  • Linear Formula C₄H₆N₂O

  • Molecular Weight 98.1 g/mol

  •  EC Number 214-013-6

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks
packaging   50 g in Glass bottle (8141820050)
mp   56 - 60 °C
pH   6.0 - 7.5 (10 g/l, H₂O, 20 °C)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 56 °C
  Melting range (upper value): ≤ 60 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2934 99 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?