• USA Home
  • 8.14183 - 4-Aminobenzonitrile

EMAIL THIS PAGE TO A FRIEND
8.14183 EMD Millipore

4-Aminobenzonitrile

4-Aminobenzonitrile for synthesis. CAS 873-74-5, chemical formula 4-(NH₂)C₆H₄CN., for synthesis

Synonym: 4-Cyanoaniline

  • CAS Number 873-74-5

  • Linear Formula C₇H₆N₂

  • Molecular Weight 118.13 g/mol

  •  EC Number 212-850-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8141830010)
mp   84 - 87 °C
LD 50 oral   LD50 Rat 283 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 84 °C
  Melting range (upper value): ≤ 87 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2926 90 70
Hazard Statement(s) 
H301: Toxic if swallowed.
Precautionary Statement(s) 
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 3439 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 3439 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 3439 , 6.1, III
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
S Phrase 
S 24/25
Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?