• USA Home
  • 8.14185 - Hydroxymethylcyclobutane

EMAIL THIS PAGE TO A FRIEND
8.14185 EMD Millipore

Hydroxymethylcyclobutane

Hydroxymethylcyclobutane for synthesis. CAS 4415-82-1, molar mass 86.13 g/mol., for synthesis

Synonym: Cyclobutylmethanol

  • CAS Number 4415-82-1

  • Linear Formula C₅H₁₀O

  • Molecular Weight 86.13 g/mol

  •  EC Number 224-575-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   142 - 144 °C (1013 hPa)
density   0.92 g/cm3 (20 °C)
flash point   40 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.919 - 0.920
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2906 19 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, III
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, III
Categories of danger 
flammable
R Phrase 
R 10
Flammable.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?