• USA Home
  • 8.14187 - 3,5-Dimethylisoxazole

EMAIL THIS PAGE TO A FRIEND
8.14187 EMD Millipore

3,5-Dimethylisoxazole

3,5-Dimethylisoxazole for synthesis. CAS 300-87-8, pH 4.5 (10 g/l, H₂O, 20 °C)., for synthesis

  • CAS Number 300-87-8

  • Linear Formula C₅H₇NO

  • Molecular Weight 97.11 g/mol

  •  EC Number 206-100-2

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks
bp   141 - 143 °C (1013 hPa)
vapor pressure   6.9 hPa (20 °C)
solubility   50.9 g/l
density   0.98 g/cm3 (20 °C)
flash point   31 °C
autoignition temp.   520 °C
mp   -14 °C
pH   4.5 (10 g/l, H₂O, 20 °C)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.982 - 0.983
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2934 99 90
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, II
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, II
Categories of danger 
flammable
R Phrase 
R 10
Flammable.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?