• USA Home
  • 8.14193 - (R)-(+)-2-Hydroxy-1,2,2-triphenylethyl acetate

EMAIL THIS PAGE TO A FRIEND
8.14193 EMD Millipore

(R)-(+)-2-Hydroxy-1,2,2-triphenylethyl acetate

(R)-(+)-2-Hydroxy-1,2,2-triphenylethyl acetate for synthesis. CAS 95061-47-5, molar mass 332.4 g/mol., for synthesis

Synonym: HYTRA, (R)-(+)-Acetic acid 2-hydroxy-1,2,2-triphenylethyl ester

  • CAS Number 95061-47-5

  • Linear Formula C₂₂H₂₀O₃

  • Molecular Weight 332.4 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 g in Glass bottle (8141930001)
mp   237 - 240 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 240 °C
  Melting range (upper value): ≤ 245 °C
  Spec. rotation [α²0/D (c=1 in pyridine): +213.0 - +216.0 °
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2915 39 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?