EMAIL THIS PAGE TO A FRIEND
8.14195 EMD Millipore

4-Phenoxyphenol

4-Phenoxyphenol for synthesis. CAS 831-82-3, molar mass 186.2 g/mol., for synthesis

Synonym: 4-Hydroxydiphenyl ether, Hydroquinone monophenyl ether

  • CAS Number 831-82-3

  • Linear Formula C₁₂H₁₀O₂

  • Molecular Weight 186.2 g/mol

  •  EC Number 212-611-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8141950010)
bp   177 - 180 °C (12 hPa)
flash point   170 °C
mp   80 - 83 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 81 °C
  Melting range (upper value): ≤ 84 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 50 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?