• USA Home
  • 8.14216 - 2,6-Dimethyl-4-nitrophenol

EMAIL THIS PAGE TO A FRIEND
8.14216 EMD Millipore

2,6-Dimethyl-4-nitrophenol

2,6-Dimethyl-4-nitrophenol for synthesis. CAS 2423-71-4, molar mass 167.16 g/mol., for synthesis

Synonym: 4-Nitro-2,6-xylenol

  • CAS Number 2423-71-4

  • Linear Formula C₈H₉NO₃

  • Molecular Weight 167.16 g/mol

  •  EC Number 219-353-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8142160005)
mp   162 - 168 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 162 °C
  Melting range (upper value): ≤ 168 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2908 99 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?