EMAIL THIS PAGE TO A FRIEND
8.14217 EMD Millipore

N-Formylurea

N-Formylurea for synthesis. CAS 1190-24-5, EC Number 214-719-4, chemical formula H₂NCONHCHO., for synthesis

  • CAS Number 1190-24-5

  • Linear Formula C₂H₄N₂O₂

  • Molecular Weight 88.06 g/mol

  •  EC Number 214-719-4

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks,
packaging   25 g in Plastic bottle (8142170025)
mp   170 - 173 °C (External MSDS)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (ex N): ≥ 99.0 %
  Melting range (lower value): ≥ 166 °C
  Melting range (upper value): ≤ 169 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2924 19 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?