• USA Home
  • 8.14221 - Benzaldehyde dimethyl acetal

EMAIL THIS PAGE TO A FRIEND
8.14221 EMD Millipore

Benzaldehyde dimethyl acetal

Benzaldehyde dimethyl acetal for synthesis. CAS No. 1125-88-8, EC Number 214-413-0., for synthesis

Synonym: α,α-Dimethoxytoluene

  • CAS Number 1125-88-8

  • Linear Formula C₉H₁₂O₂

  • Molecular Weight 152.19 g/mol

  •  EC Number 214-413-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8142210100)
bp   87 - 89 °C (24 hPa)
density   1.02 g/cm3 (20 °C)
flash point   69 °C
LD 50 oral   LD50 Rat 1220 mg/kg
LD 50 dermal   LD50 Rabbit > 5000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.015 - 1.018
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2911 00 00
Hazard Statement(s) 
H302: Harmful if swallowed.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?