• USA Home
  • 8.14228 - 3,6-Dimethylpiperazine-2,5-dione

EMAIL THIS PAGE TO A FRIEND
8.14228 EMD Millipore

3,6-Dimethylpiperazine-2,5-dione

3,6-Dimethylpiperazine-2,5-dione for synthesis. CAS 5625-46-7, molar mass 142.15 g/mol., for synthesis

Synonym: Alanine anhydride

  • CAS Number 5625-46-7

  • Linear Formula C₆H₁₀N₂O₂

  • Molecular Weight 142.15 g/mol

  •  EC Number 227-058-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Plastic bottle (8142280010)
solubility   16 g/l
mp   272 - 275 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (ex N): ≥ 98.0 %
  Melting range (lower value): ≥ 272 °C
  Melting range (upper value): ≤ 275 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 59 95
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?