• USA Home
  • 8.14234 - Methyl phenyl sulfoxide

EMAIL THIS PAGE TO A FRIEND
8.14234 EMD Millipore

Methyl phenyl sulfoxide

Methyl phenyl sulfoxide for synthesis. CAS 1193-82-4, EC Number 214-781-2, chemical formula C₆H₅SOCH₃., for synthesis

  • CAS Number 1193-82-4

  • Linear Formula C₇H₈OS

  • Molecular Weight 140.21 g/mol

  •  EC Number 214-781-2

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfoxides,
packaging   5 g in Glass bottle (8142340005)
bp   263 - 264 °C (1013 hPa)
flash point   86 °C
mp   25 - 30 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 25 °C
  Melting range (upper value): ≤ 30 °C
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2930 90 98
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?