• USA Home
  • 8.14248 - (R)-(-)-N-(3,5-Dinitrobenzoyl) phenylglycine

EMAIL THIS PAGE TO A FRIEND
8.14248 EMD Millipore

(R)-(-)-N-(3,5-Dinitrobenzoyl) phenylglycine

(R)-(-)-N-(3,5-Dinitrobenzoyl) phenylglycine for synthesis. CAS 74927-72-3, molar mass 345.26 g/mol., for synthesis

  • CAS Number 74927-72-3

  • Linear Formula C₁₅H₁₁N₃O₇

  • Molecular Weight 345.26 g/mol

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents More...
packaging   1 g in Glass bottle (8142480001)
mp   217 - 218 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 218 °C
  Melting range (upper value): ≤ 221 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2924 29 70
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?