• USA Home
  • 8.14261 - 3-Fluorobenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
8.14261 EMD Millipore

3-Fluorobenzyl alcohol

3-Fluorobenzyl alcohol for synthesis. CAS No. 456-47-3, EC Number 207-265-3., for synthesis

  • CAS Number 456-47-3

  • Linear Formula C₇H₇FO

  • Molecular Weight 126.13 g/mol

  •  EC Number 207-265-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   104 - 105 °C (29 hPa)
density   1.17 g/cm3 (20 °C)
flash point   90 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.168 - 1.170
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2906 29 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?