• USA Home
  • 8.14265 - 1,8-Dihydroxy-3,6-dithiaoctane

EMAIL THIS PAGE TO A FRIEND
8.14265 EMD Millipore

1,8-Dihydroxy-3,6-dithiaoctane

1,8-Dihydroxy-3,6-dithiaoctane for synthesis. CAS 5244-34-8, chemical formula (HOCH₂CH₂SCH₂)₂., for synthesis

Synonym: 1,2-Bis(2-hydroxyethylmercapto) ethane

  • CAS Number 5244-34-8

  • Linear Formula C₆H₁₄O₂S₂

  • Molecular Weight 182.29 g/mol

  •  EC Number 226-045-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Plastic bottle (8142650100)
bp   170 °C (1 hPa)
mp   62 - 65 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 63 °C
  Melting range (upper value): ≤ 66 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2930 90 98
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?