• USA Home
  • 8.14266 - 2-Amino-5-mercapto-1,3,4-thiadiazole

EMAIL THIS PAGE TO A FRIEND
8.14266 EMD Millipore

2-Amino-5-mercapto-1,3,4-thiadiazole

2-Amino-5-mercapto-1,3,4-thiadiazole for synthesis. CAS 2349-67-9, pH 3.9 (2 g/l, H₂O, 20 °C)., for synthesis

  • CAS Number 2349-67-9

  • Linear Formula C₂H₃N₃S₂

  • Molecular Weight 133.18 g/mol

  •  EC Number 219-078-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 g in Plastic bottle (8142660050)
solubility   2 g/l
pH   3.9 (2 g/l, H₂O, 20 °C)
bulk density   350 kg/m3
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 98.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2934 99 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?