• USA Home
  • 8.14267 - 3-Amino-5-mercapto-1,2,4-triazole

EMAIL THIS PAGE TO A FRIEND
8.14267 EMD Millipore

3-Amino-5-mercapto-1,2,4-triazole

3-Amino-5-mercapto-1,2,4-triazole for synthesis. CAS 16691-43-3, molar mass 116.14 g/mol., for synthesis

  • CAS Number 16691-43-3

  • Linear Formula C₂H₄N₄S

  • Molecular Weight 116.14 g/mol

  •  EC Number 240-735-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Plastic bottle (8142670010)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (of S): ≥ 98.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?