• USA Home
  • 8.14271 - Rhodanine-N-acetic acid

EMAIL THIS PAGE TO A FRIEND
8.14271 EMD Millipore

Rhodanine-N-acetic acid

Rhodanine-N-acetic acid for synthesis. CAS 5718-83-2, molar mass 191.21 g/mol., for synthesis

Synonym: 4-Oxo-2-thioxo-3-thiazolidinylacetic acid

  • CAS Number 5718-83-2

  • Linear Formula C₅H₅NO₃S₂

  • Molecular Weight 191.21 g/mol

  •  EC Number 227-220-1

Purchase

Properties

Related Categories Acids, Acids & Bases, Building Blocks, Chemical Synthesis, Organic Building Blocks,
packaging   25 g in Plastic bottle (8142710025)
mp   145 - 148 °C (External MSDS)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 98.0 %
  Melting range (lower value): ≥ 144 °C
  Melting range (upper value): ≤ 148 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2934 10 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?