• USA Home
  • 8.14282 - Dimethylaminotrimethylsilane

EMAIL THIS PAGE TO A FRIEND
8.14282 EMD Millipore

Dimethylaminotrimethylsilane

Dimethylaminotrimethylsilane for synthesis. CAS 2083-91-2, chemical formula (CH₃)₃SiN(CH₃)₂., for synthesis

Synonym: Dimethylaminotrimethylsilan, TMSDMA, N-Trimethylsilyldimethylamine

  • CAS Number 2083-91-2

  • Linear Formula C₅H₁₅NSi

  • Molecular Weight 117.26 g/mol

  •  EC Number 218-222-3

Purchase

Properties

bp   84 - 86 °C (1013 hPa)
vapor pressure   30 hPa (0 °C)
density   0.74 g/cm3 (20 °C)
flash point   -12 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): ≥ 97.0 %
  Density (d 20 °C/ 4 °C): 0.743 - 0.746
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 90 00
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
H314: Causes severe skin burns and eye damage.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
P403 + P235: Store in a well-ventilated place. Keep cool.
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Symbols 
Corrosive  Corrosive
Hazard Symbols 
Flammable  Flammable
Declaration (railroad and road) ADR, RID 
UN 2733 , 3 (8), II
Declaration (transport by air) IATA-DGR 
UN 2733 , 3 (8), II
Declaration (transport by sea) IMDG-Code 
UN 2733 , 3 (8), II, Segregation Group: 18 (Alkalis)
Categories of danger 
highly flammable, corrosive
R Phrase 
R 11-34
Highly flammable.Causes burns.
S Phrase 
S 9-16-26-36/37/39-45
Keep container in a well-ventilated place.Keep away from sources of ignition - No smoking.In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?