EMAIL THIS PAGE TO A FRIEND
8.14288 EMD Millipore

1-Chloroheptane

1-Chloroheptane for synthesis. CAS 629-06-1, chemical formula CH₃(CH₂)₆Cl., for synthesis

Synonym: n-Heptyl chloride

  • CAS Number 629-06-1

  • Linear Formula C₇H₁₅Cl

  • Molecular Weight 134.64 g/mol

  •  EC Number 211-070-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 ml in Glass bottle (8142880250)
bp   160 - 161 °C (1013 hPa)
density   0.87 g/cm3 (20 °C)
flash point   45 °C
autoignition temp.   220 °C
mp   -69 - -65 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.874 - 0.877
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 19 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
P262: Do not get in eyes, on skin, or on clothing.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, III
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, III
Categories of danger 
flammable
R Phrase 
R 10
Flammable.
S Phrase 
S 24
Avoid contact with skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?