• USA Home
  • 8.14289 - (Methylamino)acetaldehyde dimethyl acetal

EMAIL THIS PAGE TO A FRIEND
8.14289 EMD Millipore

(Methylamino)acetaldehyde dimethyl acetal

(Methylamino)acetaldehyde dimethyl acetal for synthesis. CAS 122-07-6, chemical formula CH₃NHCH₂CH(OCH₃)₂., for synthesis

Synonym: 2,2-Dimethoxy-N-methylethylamine

  • CAS Number 122-07-6

  • Linear Formula C₅H₁₃NO₂

  • Molecular Weight 119.16 g/mol

  •  EC Number 204-520-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8142890050)
bp   140 °C (65 hPa)
vapor pressure   8 hPa (20 °C)
density   0.92 g/cm3 (20 °C)
flash point   29 °C
autoignition temp.   260 °C
mp   -73 °C
pH   10.9 (10 g/l, H₂O, 20 °C)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.926 - 0.930
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 50 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P262: Do not get in eyes, on skin, or on clothing.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 3271 , 3, III
Declaration (transport by air) IATA-DGR 
UN 3271 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 3271 , 3, III
Categories of danger 
flammable, irritant
R Phrase 
R 10-36/37
Flammable.Irritating to eyes and respiratory system.
S Phrase 
S 24
Avoid contact with skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?