• USA Home
  • 8.14295 - Ethyl (3-aminopyrazole 4-carboxylate)

EMAIL THIS PAGE TO A FRIEND
8.14295 EMD Millipore

Ethyl (3-aminopyrazole 4-carboxylate)

Ethyl (3-aminopyrazole 4-carboxylate) for synthesis. CAS 6994-25-8, molar mass 155.16 g/mol., for synthesis

Synonym: 3-Aminopyrazole-4-carboxylic acid ethyl ester

  • CAS Number 6994-25-8

  • Linear Formula C₆H₉N₃O₂

  • Molecular Weight 155.16 g/mol

  •  EC Number 230-262-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 g in Glass bottle (8142950001)
mp   102 - 106 °C (ECHA)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): ≥ 98.0 %
  Melting range (lower value): ≥ 102 °C
  Melting range (upper value): ≤ 106 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 19 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?