• USA Home
  • 8.14314 - α-(Phenylthio)phenylacetic acid

EMAIL THIS PAGE TO A FRIEND
8.14314 EMD Millipore

α-(Phenylthio)phenylacetic acid

α-(Phenylthio)phenylacetic acid for synthesis. CAS 10490-07-0, molar mass 244.3 g/mol., for synthesis

Synonym: 2-Phenylthiophenylacetic acid

  • CAS Number 10490-07-0

  • Linear Formula C₁₄H₁₂O₂S

  • Molecular Weight 244.3 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds More...
packaging   100 g in Plastic bottle (8143140100)
mp   99 - 101 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 99.0 %
  Melting range (lower value): ≥ 99 °C
  Melting range (upper value): ≤ 101 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2930 90 98
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
4
Liquid organic acids or solutions should be diluted, if necessary, and carefully neutralized with sodium hydrogen carbonate (Cat. No. 106323) or with sodium hydroxide (Cat. No 106462). Before filling into container D, check the pH with pH universal indicator strips (Cat. No. 109535).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?