• USA Home
  • 8.14316 - Crown ether/Benzo-18-crown-6

EMAIL THIS PAGE TO A FRIEND
8.14316 EMD Millipore

Crown ether/Benzo-18-crown-6

Crown ether/Benzo-18-crown-6 for synthesis. CAS 14098-24-9, molar mass 312.37 g/mol., for synthesis

Synonym: 1,4,7,10,13,16-Hexahydroxy-16-orthocyclophane

  • CAS Number 14098-24-9

  • Linear Formula C₁₆H₂₄O₆

  • Molecular Weight 312.37 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 g in Glass bottle (8143160001)
flash point   113 °C
mp   42 - 45 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (DSC): ≥ 99.0 %
  Melting range (lower value): ≥ 42 °C
  Melting range (upper value): ≤ 46 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2932 99 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?