• USA Home
  • 8.14317 - Hexaethyltriaminophosphine

EMAIL THIS PAGE TO A FRIEND
8.14317 EMD Millipore

Hexaethyltriaminophosphine

for synthesis

Synonym: Tris(diethylamino)phosphine

  • CAS Number 2283-11-6

  • Linear Formula C₁₂H₃₀N₃P

  • Molecular Weight 247.36 g/mol

  •  EC Number 218-920-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   245 - 246 °C
density   0.89 g/cm3 (20 °C)
flash point   58 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): ≥ 98.0 %
  Density (d 20 °C/ 4 °C): 0.899 - 0.901
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 39 90
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P210: Keep away from heat.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, III
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, III
Categories of danger 
irritant
R Phrase 
R 36/37/38
Irritating to eyes, respiratory system and skin.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?