EMAIL THIS PAGE TO A FRIEND
8.14334 EMD Millipore

1,8-Dichlorooctane

1,8-Dichlorooctane for synthesis. CAS 2162-99-4, chemical formula Cl(CH₂)₈Cl., for synthesis

  • CAS Number 2162-99-4

  • Linear Formula C₈H₁₆Cl₂

  • Molecular Weight 183.12 g/mol

  •  EC Number 218-490-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8143340025)
bp   240 - 242 °C (1013 hPa)
vapor pressure   0.1 hPa (20 °C)
solubility   0.17 g/l
density   1.03 g/cm3 (20 °C)
flash point   118 °C
autoignition temp.   215 °C
mp   -8 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.025 - 1.026
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 19 00
Hazard Statement(s) 
H315: Causes skin irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 38
Irritating to skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?