• USA Home
  • 8.14335 - 4-Hydroxy-3-methoxy-5-nitrobenzaldehyde

EMAIL THIS PAGE TO A FRIEND
8.14335 EMD Millipore

4-Hydroxy-3-methoxy-5-nitrobenzaldehyde

4-Hydroxy-3-methoxy-5-nitrobenzaldehyde for synthesis. CAS 6635-20-7, molar mass 197.15 g/mol., for synthesis

Synonym: 4-Hydroxy-3-methoxy-5-nitrobenzaldehyde

  • CAS Number 6635-20-7

  • Linear Formula C₈H₇NO₅

  • Molecular Weight 197.15 g/mol

  •  EC Number 229-633-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8143350005)
flash point   >110 °C
mp   175 - 178 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 98.0 %
  Melting range (lower value): ≥ 175 °C
  Melting range (upper value): ≤ 178 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2913 00 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?