EMAIL THIS PAGE TO A FRIEND
8.14341 EMD Millipore

5-Hexen-1-ol

5-Hexen-1-ol for synthesis. CAS No. 821-41-0, EC Number 212-477-4., for synthesis

  • CAS Number 821-41-0

  • Linear Formula C₆H₁₂O

  • Molecular Weight 100.16 g/mol

  •  EC Number 212-477-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
vapor pressure   1.0 hPa (20 °C)
solubility   18.6 g/l
density   0.84 g/cm3 (20 °C)
flash point   59 °C
autoignition temp.   275 °C
mp   <-20 °C
pH   7 (H₂O)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 90.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.844 - 0.845
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2905 29 90
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H314: Causes severe skin burns and eye damage.
Precautionary Statement(s) 
P210: Keep away from heat.
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Disposal 
1
Strongly contaminated halogen-free organic solvents: container A.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Symbols 
Corrosive  Corrosive
Declaration (railroad and road) ADR, RID 
UN 2920 , 8 (3), II
Declaration (transport by air) IATA-DGR 
UN 2920 , 8 (3), II
Declaration (transport by sea) IMDG-Code 
UN 2920 , 8 (3), II
Categories of danger 
corrosive
R Phrase 
R 34
Causes burns.
S Phrase 
S 26-36/37/39-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?