• USA Home
  • 8.14357 - 3-Aminoquinuclidine dihydrochloride

EMAIL THIS PAGE TO A FRIEND
8.14357 EMD Millipore

3-Aminoquinuclidine dihydrochloride

for synthesis

  • CAS Number 6530-09-2

  • Linear Formula C₇H₁₆Cl₂N₂

  • Molecular Weight 199.12 g/mol

  •  EC Number 229-424-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8143570005)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 99.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 39 99
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?