• USA Home
  • 8.14361 - 1,1,1,5,5,5-Hexafluoroacetylacetone

EMAIL THIS PAGE TO A FRIEND
8.14361 EMD Millipore

1,1,1,5,5,5-Hexafluoroacetylacetone

1,1,1,5,5,5-Hexafluoroacetylacetone for synthesis. CAS No. 1522-22-1, EC Number 216-191-0., for synthesis

Synonym: 1,1,1,5,5,5-Hexafluoro-2,4-pentadione

  • CAS Number 1522-22-1

  • Linear Formula C₅H₂F₆O₂

  • Molecular Weight 208.06 g/mol

  •  EC Number 216-191-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8143610005)
density   1.48 - 1.49 g/cm3 (20 °C)
flash point   32 °C
LD 50 oral   LD50 Rat 290 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.487 - 1.494
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 79 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H301: Toxic if swallowed.
H314: Causes severe skin burns and eye damage.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Corrosive  Corrosive
Declaration (railroad and road) ADR, RID 
UN 2920 , 8 (3), II
Declaration (transport by air) IATA-DGR 
UN 2920 , 8 (3), II
Declaration (transport by sea) IMDG-Code 
UN 2920 , 8 (3), II
Categories of danger 
flammable, harmful, corrosive
R Phrase 
R 10-22-34-37
Flammable.Harmful if swallowed.Causes burns.Irritating to respiratory system.
S Phrase 
S 26-36/37/39-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?