EMAIL THIS PAGE TO A FRIEND
8.14432 EMD Millipore

1-Bromooctadecane

1-Bromooctadecane for synthesis. CAS 112-89-0, chemical formula CH₃-(CH₂)₁₆-CH₂-Br., for synthesis

Synonym: n-Octadecyl bromide

  • CAS Number 112-89-0

  • Linear Formula C₁₈H₃₇Br

  • Molecular Weight 333.39 g/mol

  •  EC Number 204-013-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   214 °C (20 hPa)
density   0.976 g/cm3
flash point   >112 °C
mp   24 - 27 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 24 °C
  Melting range (upper value): ≤ 27 °C
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 39 19
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?