• USA Home
  • 8.14443 - 4-Phenylthiosemicarbazide

EMAIL THIS PAGE TO A FRIEND
8.14443 EMD Millipore

4-Phenylthiosemicarbazide

4-Phenylthiosemicarbazide for synthesis. CAS 5351-69-9, molar mass 167.22 g/mol., for synthesis

  • CAS Number 5351-69-9

  • Linear Formula C₇H₉N₃S

  • Molecular Weight 167.22 g/mol

  •  EC Number 226-329-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Plastic bottle (8144430010)
mp   138 - 141 °C
shipped in   ambient
storage temp.   Store at +15°C to +25°C.
specifications   Assay (HClO₄): ≥ 98.0 %
  Melting range (lower value): ≥ 136 °C
  Melting range (upper value): ≤ 140 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2930 90 98
Storage class 
6.1A Combustible substances, toxic
WGK 
WGK 3 highly water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Declaration (railroad and road) ADR, RID 
UN 2811 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 2811 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 2811 , 6.1, II
S Phrase 
S 24/25
Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?