• USA Home
  • 8.14454 - 4-Amino-3-nitrophenol

EMAIL THIS PAGE TO A FRIEND
8.14454 EMD Millipore

4-Amino-3-nitrophenol

4-Amino-3-nitrophenol for synthesis. CAS 610-81-1, molar mass 154.12 g/mol., for synthesis

  • CAS Number 610-81-1

  • Linear Formula C₆H₆N₂O₃

  • Molecular Weight 154.12 g/mol

  •  EC Number 210-236-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8144540005)
mp   149 - 152 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): ≥ 98.0 %
  Melting range (lower value): ≥ 149 °C
  Melting range (upper value): ≤ 152 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 29 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?