• USA Home
  • 8.14456 - 4-Ethylcyclohexanone

EMAIL THIS PAGE TO A FRIEND
8.14456 EMD Millipore

4-Ethylcyclohexanone

4-Ethylcyclohexanone for synthesis. CAS 5441-51-0, molar mass 126.19 g/mol., for synthesis

  • CAS Number 5441-51-0

  • Linear Formula C₈H₁₄O

  • Molecular Weight 126.19 g/mol

  •  EC Number 226-629-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8144560025)
bp   192 - 194 °C
density   0.91 g/cm3 (20 °C)
flash point   63 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.913 - 0.915
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 29 00
Hazard Statement(s) 
H315: Causes skin irritation.
H317: May cause an allergic skin reaction.
H412: Harmful to aquatic life with long lasting effects.
Precautionary Statement(s) 
P262: Do not get in eyes, on skin, or on clothing.
P273: Avoid release to the environment.
P280: Wear protective gloves.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant, sensitizing, dangerous for the environment
R Phrase 
R 38-43-52/53
Irritating to skin.May cause sensitisation by skin contact.Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 24/25-37-61
Avoid contact with skin and eyes.Wear suitable gloves.Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?