• USA Home
  • 8.14459 - Cyclopropyl methyl chloride

EMAIL THIS PAGE TO A FRIEND
8.14459 EMD Millipore

Cyclopropyl methyl chloride

Cyclopropyl methyl chloride for synthesis. CAS 5911-08-0, molar mass 90.55 g/mol., for synthesis

Synonym: (Chloromethyl)cyclopropane

  • CAS Number 5911-08-0

  • Linear Formula C₄H₇Cl

  • Molecular Weight 90.55 g/mol

  •  EC Number 227-632-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8144590005)
bp   87 - 89 °C
density   0.98 g/cm3 (20 °C)
flash point   -1 °C
mp   -91 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 95.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.980 - 0.990
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 89 80
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P403 + P235: Store in a well-ventilated place. Keep cool.
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Flammable  Flammable
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, II
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, II
Categories of danger 
highly flammable, irritant
R Phrase 
R 11-36/37/38
Highly flammable.Irritating to eyes, respiratory system and skin.
S Phrase 
S 16
Keep away from sources of ignition - No smoking.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?