• USA Home
  • 8.14460 - 3-Amino-5-(methylmercapto)-1H-1,2,4-triazole

EMAIL THIS PAGE TO A FRIEND
8.14460 EMD Millipore

3-Amino-5-(methylmercapto)-1H-1,2,4-triazole

3-Amino-5-(methylmercapto)-1H-1,2,4-triazole for synthesis. CAS 45534-08-5, molar mass 130.17 g/mol., for synthesis

  • CAS Number 45534-08-5

  • Linear Formula C₃H₆N₄S

  • Molecular Weight 130.17 g/mol

  •  EC Number 256-242-4

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfides/Disulfides,
packaging   10 g in Glass bottle (8144600010)
mp   135 - 138 °C
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): ≥ 98.0 %
  Melting range (lower value): ≥ 135 °C
  Melting range (upper value): ≤ 138 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?