• USA Home
  • 8.14481 - α-Methylcinnamaldehyde

EMAIL THIS PAGE TO A FRIEND
8.14481 EMD Millipore

α-Methylcinnamaldehyde

α-Methylcinnamaldehyde for synthesis. CAS 101-39-3, molar mass 146.19 g/mol., for synthesis

Synonym: 2-Methyl-3-phenyl-2-propenal

  • CAS Number 101-39-3

  • Linear Formula C₁₀H₁₀O

  • Molecular Weight 146.19 g/mol

  •  EC Number 202-938-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 ml in Glass bottle (8144810250)
  50 ml in Glass bottle (8144810050)
bp   248 - 249 °C (1013 hPa)
density   1.03 g/cm3 (20 °C)
flash point   79 °C
LD 50 oral   LD50 Rat 2050 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.035 - 1.037
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2912 29 00
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?