• USA Home
  • 8.14482 - 4-Biphenylylacetic acid

EMAIL THIS PAGE TO A FRIEND
8.14482 EMD Millipore

4-Biphenylylacetic acid

4-Biphenylylacetic acid for synthesis. CAS 5728-52-9, molar mass 212.24 g/mol., for synthesis

  • CAS Number 5728-52-9

  • Linear Formula C₁₄H₁₂O₂

  • Molecular Weight 212.24 g/mol

  •  EC Number 227-233-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8144820025)
mp   160 - 163 °C
LD 50 oral   LD50 Rat 164 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 97.0 %
  Melting range (lower value): ≥ 160 °C
  Melting range (upper value): ≤ 163 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 39 90
Hazard Statement(s) 
H301: Toxic if swallowed.
Precautionary Statement(s) 
P262: Do not get in eyes, on skin, or on clothing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK 
WGK 3 highly water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 2811 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 2811 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 2811 , 6.1, III
Categories of danger 
toxic
R Phrase 
R 25
Toxic if swallowed.
S Phrase 
S 24/25-37-45
Avoid contact with skin and eyes.Wear suitable gloves.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?