• USA Home
  • 8.14493 - 2,6,6-Trimethyl-2-cyclohexene-1,4-dione

EMAIL THIS PAGE TO A FRIEND
8.14493 EMD Millipore

2,6,6-Trimethyl-2-cyclohexene-1,4-dione

2,6,6-Trimethyl-2-cyclohexene-1,4-dione for synthesis. CAS 1125-21-9, molar mass 152.19 g/mol., for synthesis

Synonym: 4-Oxoisophorone

  • CAS Number 1125-21-9

  • Linear Formula C₉H₁₂O₂

  • Molecular Weight 152.19 g/mol

  •  EC Number 214-406-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   222 °C
density   1.03 g/cm3 (20 °C)
flash point   94 °C
autoignition temp.   460 °C
mp   15 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.031 - 1.034
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 29 00
Hazard Statement(s) 
H317: May cause an allergic skin reaction.
Precautionary Statement(s) 
P280: Wear protective gloves.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
R Phrase 
R 43
May cause sensitisation by skin contact.
S Phrase 
S 24-37
Avoid contact with skin.Wear suitable gloves.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?