• USA Home
  • 8.14498 - (R)-(+)-2-Chloropropionic acid

EMAIL THIS PAGE TO A FRIEND
8.14498 EMD Millipore

(R)-(+)-2-Chloropropionic acid

for synthesis

  • CAS Number 7474-05-7

  • Linear Formula C₃H₅ClO₂

  • Molecular Weight 108.52 g/mol

  •  EC Number 209-952-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8144980005)
bp   185 °C (1013 hPa)
density   1.25 g/cm3 (20 °C)
flash point   101 °C
autoignition temp.   550 °C
mp   -12 °C
LD 50 oral   LD50 Rat 500 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.257 - 1.258
  Spec. rotation [α²0/D (undiluted): +12.0 - +14.0 °
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2915 90 70
Hazard Statement(s) 
H302: Harmful if swallowed.
H314: Causes severe skin burns and eye damage.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
8A Combustible, corrosive hazardous materials
WGK 
WGK 1 slightly water endangering
Disposal 
4
Liquid organic acids or solutions should be diluted, if necessary, and carefully neutralized with sodium hydrogen carbonate (Cat. No. 106323) or with sodium hydroxide (Cat. No 106462). Before filling into container D, check the pH with pH universal indicator strips (Cat. No. 109535).
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Corrosive  Corrosive
Declaration (railroad and road) ADR, RID 
UN 2511 , 8, III
Declaration (transport by air) IATA-DGR 
UN 2511 , 8, III
Declaration (transport by sea) IMDG-Code 
UN 2511 , 8, III, Segregation Group: 1 (Acids)
Categories of danger 
harmful, corrosive
R Phrase 
R 22-35
Harmful if swallowed.Causes severe burns.
S Phrase 
S 23-26-28-36-45
Do not breathe vapour.In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.After contact with skin, wash immediately with plenty of soap and water.Wear suitable protective clothing.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?