• USA Home
  • 8.14502 - Ethyl 2-cyano-3-ethoxyacrylate

EMAIL THIS PAGE TO A FRIEND
8.14502 EMD Millipore

Ethyl 2-cyano-3-ethoxyacrylate

Ethyl 2-cyano-3-ethoxyacrylate for synthesis. CAS 94-05-3, chemical formula C₂H₅OCHC(CN)COOC₂H₅., for synthesis

Synonym: Ethyl(ethoxymethylene)cyanoacetate, Ethoxymethylenecyanoacetic acid ethyl ester

  • CAS Number 94-05-3

  • Linear Formula C₈H₁₁NO₃

  • Molecular Weight 169.18 g/mol

  •  EC Number 202-299-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8145020025)
bp   190 - 191 °C (40 hPa)
vapor pressure   0.1 hPa (20 °C)
flash point   130 °C
mp   48 - 51 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 49 °C
  Melting range (upper value): ≤ 52 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2926 90 70
Precautionary Statement(s) 
P262: Do not get in eyes, on skin, or on clothing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
6
Sulfides and sulfites as well as other reducing substances, inorganic cyanides and isonitriles: Stir into sodium hypochlorite solution (Cat. No. 105614) and, if necessary, leave to react for several days. Caution: some substances may show a violent reaction! Draw off any toxic or flammable gases that may be formed. Excess oxidant should be neutralized with sodium thiosulfate (Cat. No. 106513). Container D or E.
S Phrase 
S 24/25
Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?