• USA Home
  • 8.14504 - 3-Furancarboxylic acid

EMAIL THIS PAGE TO A FRIEND
8.14504 EMD Millipore

3-Furancarboxylic acid

3-Furancarboxylic acid for synthesis. CAS 488-93-7, molar mass 112.08 g/mol., for synthesis

Synonym: 3-Furoic acid

  • CAS Number 488-93-7

  • Linear Formula C₅H₄O₃

  • Molecular Weight 112.08 g/mol

  •  EC Number 207-689-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
mp   119 - 122 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 97.0 %
  Melting range (lower value): ≥ 119 °C
  Melting range (upper value): ≤ 122 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2932 19 00
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
8A Combustible, corrosive hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 3261 , 8, III
Declaration (transport by air) IATA-DGR 
UN 3261 , 8, III
Declaration (transport by sea) IMDG-Code 
UN 3261 , 8, III, Segregation Group: 1 (Acids)
Categories of danger 
irritant
R Phrase 
R 36/38
Irritating to eyes and skin.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?