• USA Home
  • 8.14510 - Trimethylvinylsilane

EMAIL THIS PAGE TO A FRIEND
8.14510 EMD Millipore

Trimethylvinylsilane

Trimethylvinylsilane for synthesis. CAS 754-05-2, molar mass 100.24 g/mol., for synthesis

Synonym: Vinyltrimethyl silane

  • CAS Number 754-05-2

  • Linear Formula C₅H₁₂Si

  • Molecular Weight 100.24 g/mol

  •  EC Number 212-042-9

Purchase

Properties

packaging   10 ml in Glass bottle (8145100010)
bp   55 - 56 °C (1013 hPa)
density   0.69 g/cm3 (20 °C)
flash point   -24 °C
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.692 - 0.696
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 90 00
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P210: Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
P240: Ground/bond container and receiving equipment.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P403 + P233: Store in a well-ventilated place. Keep container tightly closed.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Flammable  Flammable
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, II
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, II
Categories of danger 
highly flammable, irritant
R Phrase 
R 11-36/38
Highly flammable.Irritating to eyes and skin.
S Phrase 
S 16
Keep away from sources of ignition - No smoking.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?